missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-1 Antibody (CL6365) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-44429
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SSEA-1 Monoclonal antibody specifically detects SSEA-1 in Human samples. It is validated for Western Blot
Spezifikation
| SSEA-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL | |
| alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
| This antibody was generated using a synthetic peptide of P22083, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells | |
| 2526 | |
| Human | |
| Purified |
| Western Blot | |
| CL6365 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur