missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SSEA5 Monoclonal Antibody (8E11-SSEA5), eBioscience™
GREENER_CHOICE

Mouse Monoclonal Antibody

279.00 €

Spezifikation

Antigen SSEA5
Klon 8E11-SSEA5
Konzentration 0.5 mg/mL
Inhalt und Lagerung 4°C
Anwendungen Flow Cytometry, Western Blot, Immunocytochemistry
Mehr Spezifikationen ansehen

Produkte 1
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15218107
GREENER_CHOICE
Unterlagen ansehen
Invitrogen™
14-8857-82
100 μg
279.00 €
100 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung

Beschreibung

Description: The monoclonal antibody 8E11-SSEA5 recognizes human stage-specific embryonic antigen-5 (SSEA-5), an H type-1 glycan. SSEA-5 belongs to the SSEA family of glycoproteins and is highly expressed on pluripotent human stem cells including embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. During embryonic development, SSEA-5 is expressed within the blastocyst on inner cell mass (ICM) cells that later give rise to ES cells. As ES and iPS cells differentiate, SSEA-5 expression is downregulated. The tumorigenic potential of contaminating stem cells in cell populations for transplantation is a hurdle to using this approach for disease therapy. The 8E11 antibody in combination with either CD9 and CD90 or CD50 and CD200 to sort populations has been shown to effectively separate teratoma-forming cells from stem cell-derived populations of differentiated cells. Applications Reported: This 8E11-SSEA5 antibody has been reported for use in flow cytometric analysis, western blotting, and immunocytochemistry. Applications Tested: This 8E11-SSEA5 antibody has been tested immunocytochemistry on unfixed human Ntera2 cells at less than or equal to 10 μg/mL. The 8E11-SSEA5 antibody has also been tested by flow cytometric analysis on NTERA-2 cells at less than or equal to 0.25 μg/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less...

Stage-Specific Embryonic Antigen-5 (SSEA-5) is an oligosaccharide that can be part of glycosphingolipids and glycoproteins. It contains the motif Fuc1-2Gal-beta1-3GlcNAc-beta, which is characteristic of H-1 antigens. SSEA-5 is found in the inner cell mass of the human blastocyst during embryogenesis, on undifferentiated human embryonic stem cells and on the surface of induced pluripotent cells. SSEA-5 was found to proceed the loss of other known stem cell markers and the combination of SSEA-5 with either CD9 and CD90 or CD50 and CD200 was superior in identifying and eliminating rare undifferentiated human ES cells from differentiated cell populations than the use of other established stem cell markers.
Spezifikation

Spezifikation

SSEA5
0.5 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Mouse
RUO
PBS with 0.09% sodium azide; pH 7.2
Primary
Affinity chromatography
8E11-SSEA5
4°C
Monoclonal
Liquid
IgG2b κ
Human
SSEA-5
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt