Learn More
Invitrogen™ SSTR2 Recombinant Rabbit Monoclonal Antibody (HL3078)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556879
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SSTR2 Recombinant Monoclonal Antibody for ICC/IF, IHC (P)
SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
Spezifikation
| SSTR2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P30874 | |
| SSTR2 | |
| Synthetic peptide encompassing a sequence within the Extracellular domain of human Somatostatin receptor 2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| HL3078 | |
| Unconjugated | |
| SSTR2 | |
| Smstr2; Smstr-2; somatostatin receptor; somatostatin receptor 2; somatostatin receptor 2a; somatostatin receptor subtype 2; somatostatin receptor type 2; somatotropin release-inhibiting factor receptor; SRIF-1; SS2R; SS-2-R; SS2-R; SSR2; sst2; Sstr2; SSTR-2; SSTR2A | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6752 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |