Learn More
Invitrogen™ SSX2IP Recombinant Rabbit Monoclonal Antibody (24GB5255)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557274
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SSX2IP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a protein that binds the cancer-testis antigen Synovial Sarcoma X breakpoint 2 protein. The encoded protein may regulate the activity of Synovial Sarcoma X breakpoint 2 protein in malignant cells. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 3.
Spezifikation
| SSX2IP | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8CGZ2, Q9Y2D8 | |
| SSX2IP | |
| A synthesized peptide derived from human SSX2IP (aa 580-614). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5255 | |
| Unconjugated | |
| SSX2IP | |
| ADIP; Afadin- and alpha-actinin-binding protein; afadin- and alpha-actinin-binding protein ADIP; Afadin- and alpha-actinin-binding protein-like protein; afadin DIL domain-interacting protein; AU014939; AU042321; FLJ10848; KIAA0923; LOW QUALITY PROTEIN: afadin- and alpha-actinin-binding protein; MGC75026; SSX family member 2 interacting protein; SSX2-interacting protein; Ssx2ip; synovial sarcoma, X breakpoint 2 interacting protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 117178, 308023 | |
| -20°C | |
| Liquid |