missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ST2 Recombinant Rabbit Monoclonal Antibody (10L0B1)
GREENER_CHOICE

Artikelnummer. 30283094
Change view
Click to view available options
Menge:
20 μL
100 μL
Unit Size:
100 Mikroliter
20 Mikroliter
Product Code. Menge unitSize
30283094 100 μL 100 Mikroliter
30282819 20 μL 20 Mikroliter
2 options
This item is not returnable. View return policy

Product Code. 30283094

Brand: Invitrogen™ MA557972

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: SKSSWGLENE ALIVRCPQRG RSTYPVEWYY SDTNESIPTQ KRNRIFVSRD RLKFLPARVE DSGIYACVIR SPNLNKTGYL NVTIHKKPPS CNIPDYLMYS TVRGSDKNFK ITCPTIDLYN WTAPVQWFKN CKALQEPRFR AHRSYLFIDN VTHDDEGDYT CQFTHAENGT NYIVTATRSF TVEEKGFSMF PVITNPPYNH TMEVEIGKPA SIACSACFGK GSHFLADVLW QINKTVVGNF GEARIQEEEG RNESSSNDMD CLTSVLRITG VTEKDLSLEY DCLALNLHGM IRHTIRLRRK QPIDHR.

IL-33R, also known as ST2, is a receptor for IL-33. It is a member of the IL-1 receptor family of innate receptors. Its extracellular domain consists of 3 Ig-like domains that directly interact with its co-receptor IL-1RAcP, and the ligand alarmin, IL-33. IL-33 ligation results in the association of IL-33R complex with its adaptor proteins MyD88, IRAK1, IRAK4 and TRAF6, and activation of the NF-kB, and MAPK pathways. This, in turn, instigates the release of chemokines and cytokines such as IL-5, IL-6, IL-8, and IL-13. Two most common isoforms of IL-33R that result from alternative splicing are ST2L - the membrane bound form, and ST2S - the soluble form. The membrane bound form can be released from the cell surface by proteolytic cleavage. The soluble forms have been suggested to act as decoys dampening the IL-33 signaling. IL-33R is primarily expressed by mast cells, basophils, and eosinophils. In mouse, IL-33R expression has been shown on ILC2 and Th2 cells. IL-33R signaling has been implicated in the number of immune conditions, including allergies, arthritis, atherosclerosis and sepsis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST2
Anwendungen ELISA, Western Blot
Klassifikation Recombinant Monoclonal
Klon 10L0B1
Konzentration 1.9 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gen IL1RL1
Gen-Zugriffsnummer P14719
Gen-Alias DER4; Fit1; Fit-1; Fos-responsive gene 1 protein; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; IL-1 R4; IL1RL1; IL-1RL1; IL33R; IL-33R; interleukin 1 receptor like 1; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin1 receptor like 1; interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Ly84; lymphocyte antigen 84; Protein ST2; Protein T1; sIL 33R; soluble IL 33 Receptor; soluble IL 33R; St2; ST2L; St2-rs1; ST2V; Ste2; T1; T1/ST2
Gensymbole IL1RL1
Wirtsspezies Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 27-332 of mouse IL-33R (ST2) (NP_001020773.1).
Reinigungsverfahren Protein A
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 17082
Zielspezies Mouse
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.