missing translation for 'onlineSavingsMsg'
Learn More

Amsta Rostfritt Stainless Steel Bottle Base Without Hooks Handle Skirtband or Lid

Product Code. 30285007
Click to view available options
Kapazität (metrisch):
1 l
1,5 L
2 l
3 l
Unit Size:
1 Stück
10 Stück
This item is not returnable. View return policy

Product Code. 30285007

Brand: Amsta Rostfritt 506031,5

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Highest quality stainless steel bottle. Manufactured in Sweden.

  • Stainless steel 304 /1.4301

Specifications

Produkttyp Pail
Kapazität (metrisch) 1,5 L
Farbe Silver
Material Stainless Steel
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.