Learn More
Cole-Parmer™ EDELSTHAL MESH SIEB 90MM 213μM 213μM
Artikelnummer. 11759443 Alle ansehen Cole-Parmer Produkte
Change view
Click to view available options
Material:
Edelstahl
Packungsgröße:
5 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Material unitSize
11759443 Edelstahl 5 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11759443 Lieferant Cole-Parmer™ Lieferanten-Nr. 0663542
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Spezifikation
Material Edelstahl
Porengröße 213 μm
Menge 5Pack
Dicke (metrisch) 178 μm
Typ Bildschirmscheibe
Abmessungen (L x B) 30 x 30cm