missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stanozolol-d3 (17-methyl-d3), CDN
Click to view available options
Menge:
0.01 g
0.05 g
Unit Size:
0.01 Gramm
0.05 Gramm
Specifications
Specifications
| Chemischer Name oder Material | Stanozolol-d3 |
| CAS | 792214-66-5 |
| Summenformel | C21 D3 H29 N2 O |
| Menge | 0.05 g |
| Synonym | 1'H-Androstano[3,2-c]pyrazol-17-ol, 17-(methyl-d3)-, (5α,17β)- (9 CI), Stanozolol D3 (17-methyl D3), Stanozolol-d3, (5α,17β)-17-(Methyl-d3)-2'H-androst-2-eno[3,2-c]pyrazol-17-ol, 17-(Methyl-d3)-5α-androstano[3,2-c]pyrazol-17β-ol, 17β-Hydroxy-17-(methyl-d3)-5α-androstano[3,2-c]pyrazole |
| InChI-Formel | InChI=1 S/C21H32N2O/c1-19-11-13-12-22-23-18(13)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,24 H,4-11H2,1-3H3,(H,22,23)/t14-,15+,16-,17-,19-,20-,21-/m0/s1/i3D3 |
| SMILES | [2 H]C([2 H])([2 H])[C@]1(O)CC[C@H]2 [C@@H]3 CC[C@H]4 Cc5[nH]ncc5C[C@]4(C)[C@H]3 CC[C@]12 C |
| Molekulargewicht (g/mol) | 331.51 |
| Formelmasse | 331.27 g/mol |
| Reinheit (%) | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction