Learn More
Stärke, löslich, ACS (für die Iodometrie), Thermo Scientific Chemicals
Artikelnummer. p-7050395
Click to view available options
Menge:
50 g
250 g
1 kg
Packungsgröße:
1 Kilogramm
250 Gramm
50 Gramm
Artikelnummer. 11394227
Marke: Thermo Scientific Alfa Aesar 036703.18
Bulk oder benutzerdefiniertes Format anfordern
om dit product te kopen Registreer vandaag om een webaccount aan te maken
For Jodometrie
In food industry, starch is used as a food preservative. Starch has extensive industrial use as an adhesive or bonding agent. It is also used in paper industry. Starch is used in the procedure to manufacture gypsum wallboards. Starch used in the laundry, also known as clothing starch. Glucose derived from starch may be fermented further to make ethanol or biofuel. Starch is also employed in the petroleum exploration industry. It is interesting to note that starch may be used to make hydrogen by making usage of enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 9005-84-9
Summenformel C12H22O11
Molekulargewicht (g/mol) 342.297
MDL-Nummer MFCD00082026
InChI-Schlüssel GUBGYTABKSRVRQ-ASMJPISFSA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC-Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
Spezifikation
Chemischer Name oder Material Starch
Namenshinweis soluble
Physikalische Form Pulver
Schmelzpunkt 256°C to 258°C (decomposition)
Hinweise zur Reinheitsqualität For iodometry
Merck Index 14,8799
Löslichkeitsinformationen Soluble in hot water.
Menge 50 g
Güte ACS