missing translation for 'onlineSavingsMsg'
Learn More

STAT3 Antibody (CL0490) [PE], Novus Biologicals™

Product Code. 30303499 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
0.1 mL
missing translation for 'unitSize'
0.10 Milliliter
Product Code. Menge unitSize
30303499 0.1 mL 0.10 Milliliter
1 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 30303499

missing translation for 'mfr': Novus Biologicals NBP252935PE

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAT3
Anwendungen Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
Klassifikation Monoclonal
Klon CL0490
Konjugat PE
Zusammensetzung PBS
Gen-Alias Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
Wirtsspezies Mouse
Immunogen This antibody was developed using a recombinant protein derived from P40763, with the exact immunogen sequence remaining proprietary.
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Cellular Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primär oder sekundär Primary
Gen-ID (Entrez) 6774
Zielspezies Human
Inhalt und Lagerung Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.