Learn More
Bochem™ Stahldrahtnetz
Artikelnummer. 10172361
Ansicht ändern
Click to view available options
Abmessungen (L x B):
120 x 120 mm
200 x 200 mm
Unit Size:
1 Stück
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Abmessungen (L x B) Unit Size
10172361 200 x 200 mm 1 Stück
10410353 120 x 120 mm 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10172361 Supplier Bochem™ Supplier No. 12753
  • ONLINE EXCLUSIVE PRICE
    14.00 €
    7.26 € / 1 Stück
    Sparen 6.74 €
    48% Rabatt
Verfügbar ab: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Produkttyp Drahtgewebe mit Keramikmitte
Keramikzentrum 135 mm
Abmessungen (L x B) 200 x 200 mm
Form Quadratisch