Learn More
Thermo Scientific Chemicals Stem-Cell Factor/c-Kit Inhibitor
Click to view available options
Menge:
10mg
50mg
Packungsgröße:
10 Milligramm
50 Milligramm
Beschreibung
Inhibits in vitro phosphorylation of c-Kit. Also inhibits SCF (stem cell factor)-induced c-kit phosphorylation in 501mel human melanoma cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Molekulargewicht | 355.50 |
| Menge | 50mg |
| Verpackung | Plastic bottle |
| Empfindlichkeit | Light sensitive |
| Form | Solid |