Learn More
Isofield™ Steridon Sterile Cleanroom Coverall with Hood and Integrated Facemask - Grade A/B Compatible
BIOPHARMAGREENER_CHOICE
Artikelnummer. p-200088563
Click to view available options
Größe:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Packungsgröße:
15 Stück
20 Stück
Artikelnummer. 30178872
Marke: Isofield™ 80231XXS
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
The Steridon coverall with facemask is aseptically folded and can be donned in 60 seconds or less, validated sterile via X-RAY, and tested against chemo to ASTM F 739. The anti-static, low linting, viral barrier provides essential protection in Grade A/B environments for all cleanroom activities.
  • Sterile Cleanroom coverall with hood and Integrated facemask
  • Latex-free, enclosed and elasticated face, wrists and ankles
  • Ergo-fit design at back and shoulders allowing greater range of motion
  • Integrated facemask with a guided tie-on system to facilitate easy donning and secure fit
  • Aseptically presented fold
  • Color coded left and right inner tabs to facilitate ease of donning
  • Removable external tabs at zipper to ensure sterile field maintained
  • Validated for sterility by X-Ray
  • Sterility Assurance Level (SAL) 10-6
Technical and Documentation Compliance
  • COA, COC, COI Available for download online
  • QR Code on cartons allows for immediate Document retrieval
  • Permeation of Liquids and Gases through Protective Clothing under conditions of Continuous Contact ASTM F739-20
  • Resistance of Materials Used in Protective Clothing to Penetration by Synthetic Blood ISO 16603:2004
  • Viral penetration test (liquid challenge: Bacteriophage Phi-X174) ISO 16604:2004
  • Material resistance to wet bacterial Penetration ISO 22610
  • Resistance to penetration by biologically contaminated aerosols ISO 22611
  • Resistance to dry microbial penetration ISO 22612
  • Basis Weight ASTM D3776
  • Dry Mullen Burst Strength TAPPI T403
  • Grab Tensile Strength CD ASTM D5034
  • Grab Tensile Strength MD ASTM D5034
  • Trapezoid Tear MD ASTM D5733-95
  • Wet Peel CD (inner/outer) ASTM F904-98
  • Hydrostatic Head (with screen) AATCC 127
  • Moisture Vapor Transmission Rate (MVTR) ASTM E96 Procedure D
  • Static Decay MD/CD WSP 40.2
  • Sterility Validation EN ISO 11137 Part 2 : 2015
  • Particulate Cleanliness – Helmke Drum Test : Class 1
  • Surface Resistivity ASTM D257
Spezifikation
Produkttyp Cleanroom Coveralls
Größe 2X-Small
Geschlecht Unisex
Material Polyethylene, Non-woven Polypropylene Laminate
Farbe White
Einwegprodukt Single-use
Taillenform Elastic at Back
Knöchelform Open
Stulpenform Covered Elastic
Flaschenhalstyp Hooded
Hood Type Attached, Open Face
Verschlusstyp Zipper
Saumtyp Bound
Sterilität Sterile
Widerstand Chemicals, Anti-static, Liquid, Particle
Anzahl Taschen None
Cleanroom Class Class 100 (ISO 5)