Learn More
Sterol carrier protein 2 Antibody (SR2036), Novus Biologicals™
Rabbit Monoclonal Antibody
Marke:  Novus Biologicals NBP3-22523-100ul
 Weitere Artikel aus diesem Sortiment
Product Code. 18669734
  • 449.00 € / 100 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Description
Description
Sterol carrier protein 2 Monoclonal antibody specifically detects Sterol carrier protein 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterol carrier protein 2
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
6342
Store at -20° C. Avoid freeze/thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunofluorescence
SR2036
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
DKFZp686C12188, DKFZp686D11188, NLTP, non-specific lipid-transfer protein, NSL-TP, Propanoyl-CoA C-acyltransferase, SCP-2, SCP-CHI, SCPX, SCP-X, sterol carrier protein 2EC 2.3.1.176, Sterol carrier protein X
A synthesized peptide derived from human Sterol carrier protein 2 (Uniprot #: P22307)
100 μg
Primary
Human, Mouse, Rat