missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STRA8 Recombinant Rabbit Monoclonal Antibody (PSH04-55)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556412
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
STRA8 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Meiosis-inducer required for the transition into meiosis for both female and male germ cells. In female germ cells, required for premeiotic DNA replication and subsequent events in meiotic prophase. During spermatogenesis, next to its role in meiotic initiation, promotes (but is not required for) spermatogonial differentiation. Can associate with DNA (possibly in an indirect manner), and in vitro can activate DNA transcription.
Spezifikation
| STRA8 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q7Z7C7 | |
| STRA8 | |
| Recombinant protein within human Stra8 aa 1-330/330. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| PSH04-55 | |
| Unconjugated | |
| STRA8 | |
| RGD1562852; stimulated by retinoic acid 8; stimulated by retinoic acid 8 homolog; stimulated by retinoic acid gene 8; stimulated by retinoic acid gene 8 homolog; stimulated by retinoic acid gene 8 protein; Stimulated by retinoic acid gene 8 protein homolog; stra8 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 346673 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur