missing translation for 'onlineSavingsMsg'
Learn More

Phenomenex™ Strata™-X-CW 96-Well Plates (Polymeric Weak Cation)

Product Code. 31749169 Shop All Phenomenex Products
Change view
Click to view available options
Säulenbettgewicht:
10 mg
30 mg
60 mg
Unit Size:
2 Stück
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Säulenbettgewicht Unit Size
31749169 30 mg 2 Stück
31748262 10 mg 2 Stück
31741603 60 mg 2 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31749169 Supplier Phenomenex™ Supplier No. 8ES035TGB
Sparen Sie bis zu 50 % bei Phenomenex
Verwenden Sie den Aktionscode 27955
View all promotions
 Eingeschränkter Artikel
Verfügbar ab: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Capitalize on cleanliness without sacrificing recovery with Strata-X polymeric sorbents for solid phase extraction (SPE). These sorbents are available with unique selectivities developed to cover a diverse spectrum of analytes and to simplify the method development process.

  • Brand: Strata-X
  • Technique: SP
  • Separation Mode: Multi-Mode
  • Phase: X-CW, Weak Cation Mixed Mode
TRUSTED_SUSTAINABILITY

Specifications

Säulenbettgewicht 30 mg
Zur Verwendung mit (Geräte) Vacuum Manifolds or Automated SPE Equipment
Partikelgröße 33 μm
Sorbens X-CW, Weak Cation Mixed Mode
Typ 96-well Plate
Menge 2 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.