missing translation for 'onlineSavingsMsg'
Learn More

Succinic-d4 Anhydride, CDN

Artikelnummer. 30322247
Change view
Click to view available options
Menge:
1 g
Unit Size:
1 Gramm
Product Code. Menge unitSize
30322247 1 g 1 Gramm
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30322247

Brand: CDN CDND24151G

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Succinic Anhydride-d4
CAS 14341-86-7
Summenformel C4 2H4 O3
Menge 1 g
InChI-Formel InChI=1 S/C4H4O3/c5-3-1-2-4(6)7-3/h1-2H2/i1D2,2D2
SMILES [2 H]C1([2 H])C(=O)OC(=O)C1([2 H])[2 H]
IUPAC-Name 3,3,4,4-tetradeuteriooxolane-2,5-dione
Molekulargewicht (g/mol) 104.1
Formelmasse 104.0412 g/mol
Reinheit (%) 98 atom % D, min 98% Chemical Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.