missing translation for 'onlineSavingsMsg'
Learn More

Sudan 3 D6 (Naphthyl D6), Dr. Ehrenstorfer

Product Code. 30321592
Change view
Click to view available options
Menge:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30321592 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321592 Supplier Dr. Ehrenstorfer Supplier No. DREC16986107
 Eingeschränkter Artikel
Verfügbar ab: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Entdecken Sie die von Dr. Ehrenstorfer zertifizierten Referenzmaterialien: Erhältlich in verschiedenen Formaten, einschließlich Mehrkomponenten-Gemische für regulatorische Zwecke, um Ihre Lebensmittel- und Umweltanalysen mit rückverfolgbarer, ISO-zertifizierter Qualität zu unterstützen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Sudan III-D6
CAS 1014689-17-8
Summenformel C22 2H6 H10 N4 O
Menge 10 mg
Synonym 2-Naphthalen-3,4,5,6,7,8-d6-ol, 1-[2-[4-(2-phenyldiazenyl)phenyl]diazenyl]-
InChI-Formel InChI=1S/C22H16N4O/c27-21-15-10-16-6-4-5-9-20(16)22(21)26-25-19-13-11-18(12-14-19)24-23-17-7-2-1-3-8-17/h1-15,27H/i4D,5D,6D,9D,10D,15D
SMILES [2H]c1c([2H])c([2H])c2c(N=Nc3ccc(cc3)N=Nc4ccccc4)c(O)c([2H])c([2H])c2c1[2H]
IUPAC-Name 3,4,5,6,7,8-Hexadeuterio-1-[(E)-[4-[(E)-phenyldiazenyl]phenyl]diazenyl]naphthalin-2-ol
Molekulargewicht (g/mol) 358.43
Formelmasse 358,1701 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.