missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfacetamid-Natrium-Monohydrat, LoGiCal
Description
Sulfacetamid-Natrium-Monohydrat
Specifications
Specifications
| Chemischer Name oder Material | Sulfacetamide sodium salt |
| CAS | 6209-17-2 |
| Bezeichnung der Analyten oder Bestandteile | Sulfacetamide sodium salt |
| Versandbedingung | Raumtemperatur |
| Summenformel | C8 H9 N2 Na O3 . H2 O |
| Zertifizierung/Konformität | ISO 9001 |
| Synonym | Sulfacetamide Sodium Monohydrate, N-[(4-Aminophenyl)sulfonyl]acetamide sodium salt hydrate (1:1:1), N-[(4-Aminophenyl)sulfonyl]acetamide monosodium salt monohydrate, N-Sulfanilylacetamide monosodium salt monohydrate, N1-Acetylsulfanilamide sodium monohydrate, Sodium N-sulfanilylacetamide monohydrate, Sodium sulfacetamide monohydrate, Sulamyd sodium monohydrate, Sulfacetamide sodium monohydrate |
| SMILES | O.[Na+].CC(=O)[N-]S(=O)(=O)c1ccc(N)cc1 |
| Industry Type | Forensik |
| Molekulargewicht (g/mol) | 236.22 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?