missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethazin-d4, TRC

Product Code. 30426346
Change view
Click to view available options
Menge:
1 mg
Unit Size:
1 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30426346 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30426346 Supplier Toronto Research Chemicals Supplier No. TRCS6990721MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Sulfadimidine-d4
CAS 1020719-82-7
Hinweise zur Reinheitsqualität HPLC
Summenformel C12 2H4 H10 N4 O2 S
Menge 1 mg
Synonym Benzene-2,3,5,6-d4-sulfonamide','4-amino-N-(4,6-dimethyl-2-pyrimidinyl)-, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzene-2,3,5,6-d4-sulfonamide, Sulfamethazine-d4, Sulfadimidine-d4, Sulfadimidine (phenyl-D4)
InChI-Formel InChI=1S/C12H14N4O2S/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3,(H,14,15,16)/i3D,4D,5D,6D
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)Nc2nc(C)cc(C)n2
IUPAC-Name 4-Amino-2,3,5,6-tetradeuterio-N-(4,6-dimethylpyrimidin-2-yl)benzolsulfonamid
Molekulargewicht (g/mol) 282.35
Formelmasse 282.1089
Reinheit (%) >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.