Learn More
Heinz Herenz Superfrost Slides Cut Edges
Artikelnummer. 30158345
Click to view available options
:
With Plain Corner Style
Artikelnummer. 30158345
Marke: Heinz Herenz 1041002
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Superfrost slides with cut edges and cellophane.
We offer a very wide range of slides, so you are sure to find what you are looking for.
  • Within the product lines you will find cut and ground slides.
  • Cut slides are preferred for routine microscopy and offer great flexibility.
  • Ground slides prevent cuts due to the ground edges.
  • If you want to label and categorize your slides, it is best to choose slides with matted edges or a coloured field.
  • You also have the choice between slides with and without cellophane.
Spezifikation
Edges Cut
Frosted SuperFrost
Material Glass
Precleaned No
Typ Microscope Slide
Wellanzahl 0
Farbe Transparent
Art der Ecken Plain