missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript V VILO Master Mix, Primer-Free, with gDNA Removal

Artikelnummer. 32259989
Ansicht ändern
Click to view available options
Anzahl Reaktionen:
1250 Reactions
2000 Reactions
50 Reactions
500 Reactions
Enthält:
SuperScript V VILO Master Mix (Primer-Free), No RT Control, Water
SuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water
SuperScript V VILO Master Mix (Primer-free) only
Packungsgröße:
1 Stück
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Anzahl Reaktionen Enthält Packungsgröße
32259989 1250 Reactions SuperScript V VILO Master Mix (Primer-free) only 1 Stück
32260452 50 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, Water 1 Stück
32259996 500 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, Water 1 Stück
32260516 2000 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, Water 1 Stück
32260216 50 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water 1 Stück
32260139 500 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water 1 Stück
32260514 2000 Reactions SuperScript V VILO Master Mix (Primer-Free), No RT Control, ezDNase Master Mix, Water 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Artikelnummer. 32259989 Lieferant Invitrogen™ Lieferanten-Nr. 11900004
 Eingeschränkter Artikel
Verfügbar ab: 11-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Invitrogen™ SuperScript™ V VILO™ Master Mix, Primer-Free is a ready-to-use master mix designed for fast, sensitive, and reproducible cDNA synthesis with the flexibility to select your preferred priming strategy.

Invitrogen™ SuperScript™ V VILO™ Master Mix, Primer-Free is a ready-to-use master mix designed for fast, sensitive, and reproducible cDNA synthesis with the flexibility to select your preferred priming strategy. Powered by SuperScript™ V Reverse Transcriptase, the optimized master mix combines robust performance across a broad range of RNA samples with simplified reaction setup, enhanced reagent stability, and greater workflow flexibility. SuperScript V VILO Master Mix, Primer-Free is also available with ezDNase™ Master Mix for fast and convenient genomic DNA removal prior to reverse transcription.

Features of SuperScript V VILO Master Mix, Primer-Free include:

  • Broad RNA input range—supports sensitive cDNA synthesis from 0.1 pg to 5 μg of total RNA
  • Robust performance with challenging RNA—efficient cDNA synthesis from highly structured, GC-rich, degraded and inhibitor-containing RNA
  • Fast cDNA synthesis—complete reverse transcription in as little as 1-10 minutes
  • Enhanced reagent stability—supports ambient shipping and room-temperature storage for up to 1 month
  • Simple, flexible workflow—ready-to-use format simplifies reaction setup, while room-temperature assembly and extended stability of assembled reactions enable greater workflow flexibility

SuperScript V VILO Master Mix, Primer-Free combines SuperScript V RT, RNase inhibitor, buffer, and dNTPs in a single formulation while allowing users to select and optimize primers for their specific application. This flexibility supports gene-specific primers, oligo(dT), random primers, or customized primer combinations while maintaining the convenience of a ready-to-use master mix.

SuperScript V VILO Master Mix, Primer-Free with gDNA Removal allows streamlined genomic DNA removal step. The dsDNA-specific ezDNase enzyme removes contaminating genomic DNA in as little as 2 minutes without affecting RNA.

SuperScript V VILO Master Mix, Primer-Free also features a DTT-free, animal origin–free formulation for improved sustainability and simplified reagent handling.

TRUSTED_SUSTAINABILITY

Spezifikation

Konzentration 5X
Inhalt und Lagerung • SuperScript V VILO Master Mix, Primer-Free (5 mL)

Store all contents at:
• -20°C for long-term storage until expiry date
• +5°C for up to 5 months
• Room temperature for up to 1 month
Nachweisverfahren qPCR, NGS, PCR
Form Liquid
Format Master Mix
GC-Rich PCR Performance High
PCR-Methode 2-step RT-qPCR, RT-PCR
Reaktionsgeschwindigkeit 10 min.
Verfahren Reverse Transcription
Marker oder Farbstoff Not Applicable
Optimale Reaktionstemperatur 50°C
Produkttyp Reverse Transcription Reagent
Reverse Transkriptase SuperScript V
Ribonuklease-H-Aktivität Reduced
Empfindlichkeit 0.1 pg total RNA
Versandbedingung Dry Ice, Wet Ice, Ambient
Zur Verwendung mit (Anwendung) Gene Expression, Real Time PCR (qPCR), RT-PCR, cDNA Synthesis, Next-Generation Sequencing (NGS)
Thermostabilität Up to 65°
Endprodukttyp First-Strand cDNA
Anzahl Reaktionen 1250 Reactions
Reagenztyp Reverse Transcription
Größe (Endprodukt) Up to 20 kb
Ausgangsmaterial RNA
Enthält SuperScript V VILO Master Mix (Primer-free) only
Inhibitor Tolerance (Simple) High
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.