missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Survivin Antibody (32.1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB500-237F
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Survivin Monoclonal specifically detects Survivin in Human, Hamster samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
| Survivin | |
| Monoclonal | |
| FITC | |
| O15392 | |
| BIRC5 | |
| This Survivin Antibody (32.1) [FITC] was developed against full length recombinant human Survivin [UniProt# O15392] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Survivin Antibody (32.1) [NB500-237] is specific for the nuclear form of survivin. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 32.1 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
| Mouse | |
| 16 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells | |
| 332 | |
| Human, Hamster | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur