missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sutimlimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558458
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Sutimlimab is an anti-C1s antibody. Studies have shown that it can significantly increase hemoglobin levels in patients with primary cold agglutinin disease. Cold agglutinin disease (CAD) is a type of acquired autoimmune hemolytic anemia (AIHA). In cold environments, cold parts of the body will have clinical symptoms related to red blood cell aggregation (for example, the Internet Plaque or cyanosis) and hemolytic anemia.
Spezifikation
| Sutimlimab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BIVV009; IPN-009; TNT009 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur