Learn More
Invitrogen™ Sutimlimab Recombinant Monoclonal Antibody
GREENER_CHOICE
Artikelnummer. 30284044
Change view
Click to view available options
Menge:
1 mg
100 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
Artikelnummer. Menge unitSize
30284044 100 μg 100 Mikrogramm
30283402 1 mg 1 Milligramm
2 options
Artikelnummer. 30284044
Marke: Invitrogen™ MA558457
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Sutimlimab is an anti-C1s antibody. Studies have shown that it can significantly increase hemoglobin levels in patients with primary cold agglutinin disease. Cold agglutinin disease (CAD) is a type of acquired autoimmune hemolytic anemia (AIHA). In cold environments, cold parts of the body will have clinical symptoms related to red blood cell aggregation (for example, the Internet Plaque or cyanosis) and hemolytic anemia.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Sutimlimab Humanized
Anwendungen ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Klassifikation Recombinant Monoclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Gen-Alias BIVV009; IPN-009; TNT009
Wirtsspezies Human
Reinigungsverfahren Protein A
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Lyophilized
Isotype IgG4SP