missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUZ12 Monoclonal Antibody (K.6.9)

Rabbit Monoclonal Antibody
Marke: Invitrogen™ MA511188
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
It is not recommended to aliquot this antibody.
SUZ12 (polycomb protein SUZ12) is a zinc finger gene and has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. SUZ12 is also a component of the PRC2/EED-EZH2 complex which methylates Lys-9 (H3K9me) and Lys-27 (H3K27me) of histone H3 leading to transcriptional repression of the affected target gene. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region.
Spezifikation
| SUZ12 | |
| Monoclonal | |
| 84 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q15022, Q80U70 | |
| SUZ12 | |
| Synthetic peptide corresponding to the sequence of the human SUZ12 protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
| ChIP Assay, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| K.6.9 | |
| Unconjugated | |
| SUZ12 | |
| 2610028O16Rik; AI195385; AU016842; AW536442; chET 9 protein; CHET9; chromatin precipitated E2F target 9 protein; D11Ertd530e; JJAZ1; Joined to JAZF1 protein; Kiaa0160; mKIAA0160; Polycomb protein Suz12; suppressor of zeste 12 homolog; suppressor of zeste 12 homolog (Drosophila); suppressor of zeste 12 protein homolog; Suz12; SUZ12 polycomb repressive complex 2 subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23512, 52615, 688041 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur