missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syndecan-1/CD138 Antibody (DL-101), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP1-43351-0.1MG
This item is not returnable.
View return policy
Description
Syndecan-1/CD138 Monoclonal specifically detects Syndecan-1/CD138 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Syndecan-1/CD138 | |
| Monoclonal | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| SDC1 | |
| The immunogen for this antibody was CD138. | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| DL-101 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| CD138, CD138 antigen, SDC, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 6382 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction