Learn More
Syntaxin 16 Mouse anti-Human, CoraLite 594, Clone: 4A9D8, Proteintech
Mouse Monoclonal Antibody
Marke:  Proteintech CL594-66775-100UL
Artikelnummer. 16831076
  • 450.00 € / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
The Syntaxin 16 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Syntaxin 16. This antibody recognizes human antigen. The Syntaxin 16 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spezifikation
Spezifikation
Syntaxin 16
Monoclonal
2000 μg/mL
Mouse
Primary
Human
Immunofluorescence
4A9D8
CoraLite 594
100 μL
8675