Learn More
Syntenin-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 22399-1-AP-20UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16811925
  • 145.00 € / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
The protein encoded by this gene was initially identified as a molecule linking syndecan-mediated signaling to the cytoskeleton. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. This protein may also affect cytoskeletal-membrane organization, cell adhesion, protein trafficking, and the activation of transcription factors.
Spezifikation
Spezifikation
Syntenin-1
Polyclonal
Unconjugated
SDCBP
MDA 9, MDA9, Scaffold protein Pbp1, SDCBP, ST1, SYCL, Syndecan binding protein 1, Syntenin-1, TACIP18
Rabbit
Antigen Affinity Chromatography
RUO
6386
-20°C
Liquid
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00560
SDCBP
Syntenin-1 Fusion Protein Ag18078
20 μL
Primary
Human
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen