Learn More
Invitrogen™ T-bet Recombinant Rabbit Monoclonal Antibody (9M7X6)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558223
Beschreibung
Immunogen sequence: KGFRENFESM YTSVDTSIPS PPGPNCQFLG GDHYSPLLPN QYPVPSRFYP DLPGQAKDVV PQAYWLGAPR DHSYEAEFRA VSMKPAFLPS APGPTMSYYR GQEVLAPGAG WPVAPQYPPK MGPASWFRPM RTLPMEPGPG GSEGRGPEDQ GPPLVWTEIA PIRPESSDSG LGEGDSKRRR VSPYPSSGDS SSPAGAPSPF DKEAEGQFYN YFPN.
T-bet, also known as TBX21, is a T-cell specific transcription factor containing a single T-box DNA-binding domain. It controls the expression of the TH1 cytokine, interferon-gamma. Both Th1 and Th17 cells are crucial in immune regulation and autoimmune disease development. T-bet initiates Th1 lineage development from naive Th precursor cells and directs T cell differentiation to Th1 versus Th17. T-bet is cooperating with Stat4 (signal tranducer and activator of transcription 4) in programming chromatin architecture for Th1 gene expression. Genetic variations in T-bet are associated with susceptibility to aspirin-induced asthma and nasal polyps.
Spezifikation
| T-bet | |
| Recombinant Monoclonal | |
| 1.7 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9UL17 | |
| Tbx21 | |
| Recombinant protein (or fragment). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot | |
| 9M7X6 | |
| Unconjugated | |
| Tbx21 | |
| HGNC:11599; LOW QUALITY PROTEIN: T-box transcription factor TBX21; Tbet; T-bet; TBLYM; T-box 21; T-box expressed in T cells; T-box protein 21; T-box transcription factor 21; T-box transcription factor TBX21; TBT1; TBX21; T-cell-specific T-box transcription factor T-bet; T-PET; transcription factor TBLYM; transcription factor T-box 21 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 30009 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.