Learn More
Tuttnauer T-Lab Eco Vertical Autoclave
Artikelnummer. 17837628 Alle ansehen Tuttnauer Produkte
Click to view available options
Kapazität (metrisch):
60 l
85 l
Packungsgröße:
1 Stück
Artikelnummer. 17837628
Marke: Tuttnauer TLABV85230
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
This reliable workhorse is ideal for the general laboratory in need of easy operation, safety and cycle features for day-to-day needs. Intuitive, user friendly technology, including color touchscreen and multiple programmes for sterilization.
T-Lab Eco:
Affordable Performance and Trusted Reliability for your laboratory sterilization needs. T-Lab Eco was developed especially for educational institutions and research facilities. Designed with simplicity and economy in mind. Featuring the only IoT software designed for the smart laboratory, connecting sterilization devices and communicating on-site or remotely. Cloud based and paperless. This vertical floor-standing autoclave covers the fundamental needs for general lab sterilization with the aim of increasing the productivity of your laboratory. Offering universal capacity, together with the optimization of resources such as water, power and operating time results. Intended for the sterilization of a wide range of liquids and solids such as glassware, metal utensils and other laboratory items.
  • Reliable Performance - Solid, glassware and liquid loads. Efficient sterilization time including cooling.
  • Affordable - Built with just the essential features. Less water wasted, less electricity consumed, less time spent on maintenance. Cylindrical chamber is less expensive and more reliable over a long working life.
  • Plug and Play Installation - This autoclave does not require dedicated installation connections, simply a power source.
  • Versatile for all your basic needs - Sterilizes liquids, solids, sealed and unsealed bottles. Performance and quality within a small space.
  • Unparalleled User Experience - High resolution multicolour touchscreen which adjustable screen colours. Monitor your autoclave connected to a network with R.PC.R. software.
  • Standard Features - Two PT100 sensors. Built-in cooling fan. Built-in memory: 1000 cycles.
Spezifikation
Kapazität (angloamerikanisch) 18.7 gal.
Kapazität (metrisch) 85 l
Tiefe (metrisch) Kammer 70 cm
Durchmesser (angloamerikanisch) Kammer 15.6 in.
Tiefe (angloamerikanisch) Kammer 28 in.
Durchmesser (metrisch) Kammer 39 cm
Länge (angloamerikanisch) 32.4 in.
Länge (metrisch) 81 cm
Produkttyp Autoclave, Top-loading
Temperatur (angloamerikanisch) 221°F to 278°F
Temperatur (metrisch) 105°C to 137°C
Eigenschaften IOT Ready, Plug and Play