missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T4 Polynucleotide Kinase (T4 PNK) PROMO

T4 PNK catalyzes the transfer of the gamma-phosphate from ATP to the 5'-OH group of single- and double-stranded DNAs and RNAs, oligonucleotides or nucleoside 3'-monophosphates.

Marke:  Thermo Scientific™ EK0031-RMD123

Artikelnummer. 15669444

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen »

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

  • Active in restriction enzyme, RT and T4 DNA Ligase buffers.
  • Source: E. coli cells with a cloned pseT gene of bacteriophage T4
  • Molecular Weight: The enzyme is a homotetramer. It consists of four identical subunits of 28.9 kDa
Quality Control:
  • The absence of endo-, exodeoxyribonucleases and ribonucleases confirmed by appropriate quality tests. Functionally tested for labeling 5ft.-termini of DNA
Source:
  • E.coli cells with a cloned pseT gene of bacteriophage T4
Molecular Weight:
  • The enzyme is a homotetramer. It consists of four identical subunits of 28.9kDa
Definition of Activity Unit:
  • One unit of the enzyme transfers 1nmol of gamma-phosphate from ATP to 5ft.-OH DNA in 30 min. at 37°C
  • Enzyme activity is assayed in the following mixture: 100mM Tris-HCl (pH 8.0), 10mM MgCl2, 5mM DTT, 0.5mM 5ft.-OH DNA, 0.05mM ATP and 0.1MBq/ml [gamma-33P]-ATP
Storage Buffer:
  • The enzyme is supplied in: 20mM Tris-HCl (pH 7.5), 25mM KCl, 0.1mM EDTA, 2mM DTT and 50% (v/v) glycerol
  • 10X Reaction Buffer A (for forward reaction): 500mM Tris-HCl (pH 7.6 at 25°C), 100mM MgCl2, 50mM DTT, 1mM spermidine
  • 10X Reaction Buffer B (for exchange reaction): 500mM imidazole-HCl (pH 6.4 at 25°C), 180mM MgCl2, 50mM DTT, 1mM spermidine and 1mM ADP
  • 24% PEG Solution: 24% (w/v) polyethylene glycol 6000
  • Inhibition and Inactivation:
    • Inhibitors: metal chelators, phosphate and ammonium ions, KCl and NaCl at a concentration higher than 50mM
    • Inactivated by heating at 75°C for 10 min. or by addition of EDTA

    Recommended for:

    Labeling 5ft.-termini of nucleic acids (3, 4) to be used as probes for hybridization, probes for transcript mapping, markers for gel-electrophoresis, primers for DNA sequencing, primers for PCR; 5ft.-phosphorylation of oligonucleotide, PCR products, other DNA or RNA prior to ligation; Phosphorylation of PCR primers; Detection of DNA modification by the [32P]-postlabeling assay (5, 6); Removal of 3ft.-phosphate groups (2)

    Note:

    Polyethylene glycol (PEG) and spermidine improve the rate and efficiency of the phosphorylation reaction (7). PEG is used in the exchange reaction mixture. As T4 Polynucleotide Kinase is inhibited by ammonium ions, use sodium acetate to precipitate DNA prior to phosphorylation (1, 2).

    TRUSTED_SUSTAINABILITY
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Aktionen

Aktionen

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt