Learn More
Invitrogen™ TACC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA595406
Beschreibung
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: mouse testis tissue, U87 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This locus may represent a breast cancer candidate gene. It is located close to FGFR1 on a region of chromosome 8 that is amplified in some breast cancers. Three transcript variants encoding different isoforms have been found for this gene.
Spezifikation
| TACC1 | |
| Polyclonal | |
| Unconjugated | |
| TACC1 | |
| 4833447E04Rik; AA960202; B230378H13Rik; Ga55; gastric cancer antigen Ga55; KIAA1103; Tacc1; Tacc1a; Taxin-1; transforming acidic coiled-coil containing protein 1; transforming acidic coiled-coil-containing protein 1; transforming, acidic coiled-coil containing protein 1; transforming, acidic coiled-coil containing protein 1a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 320165, 6867 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| O75410, Q6Y685 | |
| TACC1 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human TACC1 (196-235aa MTEGSMGVTLEASAEADLKAGNSCPELVPSRRSKLRKPKP). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.