Learn More
Invitrogen™ Tadocizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA559797
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Tadocizumab is a biosimilar that targets ITGA2B & ITGB3. CD41 is a transmembrane glycoprotein (integrin family) composed of two chains GpiIb alpha (heavy chain; 120 kDa) and GpiIb beta (light chain; 23 kDa). CD61 non-covalently associates with CD41.
Spezifikation
| Tadocizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| C4G1; YM-337 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |