missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAF1C Recombinant Rabbit Monoclonal Antibody (3O8T1)

Artikelnummer. p-200111628
Change view
Click to view available options
Menge:
20 μL
100 μL
Packungsgröße:
100 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
30283570 20 μL 20 Mikroliter
30283859 100 μL 100 Mikroliter
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30283570

Marke: Invitrogen™ MA557997

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: PWDPGLTARD LLFRGGCRYR KRPRVVLDVT EQISRFLLDH GDVAFAPLGK LMLENFKLEG AGSRTKKKTV VSVKKLLQDL GGHQPWGCPW AYLSNRQRRF SILGGPILGT SVASHLAELL HEELVLRWEQ LLLDEACTGG ALAWVPG.

Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. TAF1C is the largest SL1-specific TAF. Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the largest SL1-specific TAF. Two transcripts encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen TAF1C
Anwendungen ELISA, Western Blot
Klassifikation Recombinant Monoclonal
Klon 3O8T1
Konzentration 1.04 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gen TAF1C
Gen-Zugriffsnummer Q15572, Q6P773
Gen-Alias MGC:39976; mTAFI95; RNA polymerase I-specific TBP-associated factor 110 kDa; RNA polymerase I-specific TBP-associated factor 95 kDa; SL1; SL1, 110kD subunit; TAF1C; TAFI110; TAFI95; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, C; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, C; transcription factor TAFI95; TATA box binding protein associated factor 1C; TATA box-binding protein-associated factor 1C; TATA box-binding protein-associated factor RNA polymerase I subunit C; TATA box-binding protein-associated factor, RNA polymerase I, subunit C; TATA-box binding protein associated factor, RNA polymerase I subunit C; TATA-box binding protein associated factor, RNA polymerase I, C; TBP-associated factor 1C; TBP-associated factor RNA polymerase I 95 kDa; transcription factor SL1; transcription factor TAFI95; transcription initiation factor SL1/TIF-IB subunit C
Gensymbole TAF1C
Wirtsspezies Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 75-221 of human TAF1C (NP_005670.4).
Reinigungsverfahren Protein A
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 361420, 9013
Zielspezies Human, Rat
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.