missing translation for 'onlineSavingsMsg'
Learn More

Taranabant ((1R,2R)stereoisomer), MedChemExpress

Artikelnummer. 16445537
Encompass_Preferred
Change view
Click to view available options
Menge:
2 mg
5 mg
10 mg
Unit Size:
10 Milligramm
2 Milligramm
5 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
16445537 5 mg 5 Milligramm
16402798 10 mg 10 Milligramm
16400006 2 mg 2 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16445537 Supplier Medchem Express Supplier No. HY10013B

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

MedChemExpress Taranabant (1R,2R)stereoisomer is the R-enantiomer of Taranabant. Taranabant is a highly potent and selective cannabinoid 1 (CB1) receptor inverse agonist.

Taranabant (1R,2R)stereoisomer is the R-enantiomer of Taranabant. Taranabant is a highly potent and selective cannabinoid 1 (CB1) receptor inverse agonist.

Specifications

Chemischer Name oder Material Taranabant ((1R,2R)stereoisomer)
Physikalische Form Solid
CAS 701977-08-4
Farbe Off-White
Menge 5 mg
Hinweise zur Reinheitsqualität Research
Summenformel C27H25ClF3N3O2
Synonym MK0364 (1R,2R)stereoisomer
Löslichkeitsinformationen DMSO : 100 mg/mL (193.82 mM; Need ultrasonic)
SMILES CC(OC1=NC=C(C(F)(F)F)C=C1)(C)C(N[C@H](C)[C@@H](C2=CC=CC(C#N)=C2)CC3=CC=C(Cl)C=C3)=O
Molekulargewicht (g/mol) 515.95
Formelmasse 515.95
Reinheit (%) 98.15%
Güte Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.