missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBX18 Monoclonal Antibody (635303)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561654
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
The T-box (Tbx) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The Tbx gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. Tbx genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in Tbx genes are associated with the onset of several human diseases. Our understanding of functional mechanisms of Tbx products has come mainly from the prototypical T/Brachyury, which is a transcription activator. The Tbx genes constitute a family of transcriptional regulatory genes that are implicated in a variety of developmental processes ranging from the formation of germ layers to the organizational patterning of the central nervous system.
Spezifikation
| TBX18 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O95935 | |
| TBX18 | |
| E. coli-derived recombinant human TBX18, Ala384-Ser504. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 635303 | |
| Unconjugated | |
| TBX18 | |
| 2810012F10Rik; 2810404D13Rik; CAKUT2; LOW QUALITY PROTEIN: T-box transcription factor TBX18; T-box 18; T-box protein 14; T-box protein 15; T-box protein 18; T-box transcription factor 18; T-box transcription factor TBX18; T-box18; TBX14; TBX15; Tbx18 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 9096 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur