missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCAP Recombinant Rabbit Monoclonal Antibody (7E2Z4)

Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Spezifikation
| Antigen | TCAP |
|---|---|
| Klon | 7E2Z4 |
| Konzentration | 1.1 mg/mL |
| Inhalt und Lagerung | -20°C, Avoid Freeze/Thaw Cycles |
| Anwendungen | ELISA, Western Blot |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30284652
|
Invitrogen™
MA558055 |
20 μL |
176.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
30284363
![]() |
Invitrogen™
MA558056 |
100 μL |
436.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Immunogen sequence: MATSELSCEV SEENCERREA FWAEWKDLTL STRPEEGCSL HEEDTQRHET YHQQGQCQVL VQRSPWLMMR MGILGRGLQE YQLPYQRVLP LPIFTPAKMG ATKEEREDTP IQLQELLALE TALGGQCVDR QEVAEITKQL PPVVPVSKPG ALRRSLSRSM SQEAQRG.
Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. TCAP is a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in TCAP gene are associated with limb-girdle muscular dystrophy type 2G.Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G.Spezifikation
| TCAP | |
| 1.1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O15273, O70548 | |
| 21393, 8557 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-167 of human TCAP (NP_003664.1). | |
| Antibody |
| 7E2Z4 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| Tcap | |
| 19 kDa sarcomeric protein; AI503981; AJ223855; CMD1N; CMH25; LGMD2G; limb girdle muscular dystrophy 2G (autosomal recessive); Tcap; T-cap; TELE; telethonin; teneurin C-terminal associated peptide; titin cap protein; titin-cap; titin-cap (telethonin) | |
| Tcap | |
| Primary | |
| Protein A |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts