missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCP1 Recombinant Rabbit Monoclonal Antibody (24GB4245)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556943
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TCP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The T Complex Polypeptide 1 (TCP-1) is approximately 60 kDa protein constitutively expressed in almost all eukaryotic cells, and is upregulated during spermatogenesis. It is found in the cytosol as a subunit of a hetero-oligomeric chaperone that is known to be involved in the folding of actin and tubulin. The family of proteins termed chaperonins act to recognize and stabilize polypeptide intermediates during folding, assembly and disassembly, and share many characteristics with Heat Shock Protein 70 (HSP70) including high abundance, induction by environmental stress, and ATPase activity. The chaperonin family includes the mitochondrial HSP60, Escherichia coli GroEL, the plastid Rubisco-subunit binding protein, and the archaebacterial protein TF55. The TCP-1 sequence shows nearly 40% identity to TF55, but only minimal similarity to HSP60 and GroEL.
Spezifikation
| TCP1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TCP1 | |
| 65 kDa antigen; AI528772; c-cpn; CCT; Cct1; Ccta; CCTalpha; CCT-alpha; chaperonin-containing T-complex alpha subunit TCP1; chaperonin-containing T-complex polypeptide alpha subunit; cytosolic chaperonin containing t-complex polypeptide 1; D6S230E; hypothetical protein; p63; tailless complex polypeptide 1; tailless complex polypeptide 1A; Tailless complex polypeptide 1B; t-complex 1; t-complex polypeptide 1; t-complex protein 1; T-complex protein 1 alpha subunit-like protein; T-complex protein 1 subunit alpha; T-complex protein 1 subunit alpha A; T-complex protein 1 subunit alpha B; T-complex protein 1, alpha subunit; Tcp1; Tcp-1; TCP-1-A; TCP-1-alpha; TCP-1-B; Tp63; TRic; Unknown (protein for MGC:133746); YD8142.13; YD8142B.04; YDR212W | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6950 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4245 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17987 | |
| TCP1 | |
| A synthesized peptide derived from human TCP1 alpha (aa 1-55). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur