missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCR V beta 5.3 Monoclonal Antibody (MEM-262)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA544006
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody recognizes an extracellular epitope on beta chains of the TCR expressed by HPB-ALL cell line [carrying V(beta5.3)] and a small subset of peripheral blood T cells. This subset is larger than that recognized by other V(beta5.3)-specific antibodies.
The TCR V beta 5.3 antibody recognizes beta chains of the TCR expressed by HPB-ALL cell line carrying V beta 5.3 and a small subset of peripheral blood T cells. This subset is larger than that recognized by other V (beta5.3) specific antibodies.
Spezifikation
| TCR V beta 5.3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P0DSE2 | |
| TRB | |
| Human thymoma cell line HPB-ALL. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| MEM-262 | |
| Unconjugated | |
| TRB | |
| T cell receptor beta locus; TCRB; TRB; TRB@ | |
| Mouse | |
| Protein A | |
| RUO | |
| 6957 | |
| 4°C, do not freeze | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur