missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Tryptophan 2, 3-dioxygenase (EC 1.13.11.11) plays a role in catalyzing the first and rat-limiting step in the kynurenine pathway, the major pathway of tryptophan metabolism.
Spezifikation
Spezifikation
| Antigen | TDO2 |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.3 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gen | TDO2 |
| Gen-Zugriffsnummer | P21643, P48775, P48776 |
| Gen-Alias | TDO, TDO2, TO, TPH2, TRPO, Tryptamin 2,3 dioxygenase, tryptophan 2,3 dioxygenase, Tryptophan oxygenase, Tryptophan pyrrolase, Tryptophanase |
| Gensymbole | Tdo2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?