missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Teriflunomide-d4, TRC
Specifications
Specifications
| Chemischer Name oder Material | A77 1726-d4 |
| CAS | 1185240-22-5 |
| Hinweise zur Reinheitsqualität | HPLC |
| Summenformel | C12 2H4 H5 F3 N2 O2 |
| Menge | 1 mg |
| Synonym | 2-Cyano-3-hydroxy-N-[2,3,5,6-tetradeuterio-4-(trifluoromethyl)phenyl]but-2-enamide, A77 1726-d4, (EZ)-Teriflunomide-d4 (phenyl-d4) |
| InChI-Formel | InChI=1S/C12H9F3N2O2/c1-7(18)10(6-16)11(19)17-9-4-2-8(3-5-9)12(13,14)15/h2-5,18H,1H3,(H,17,19)/i2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)C(=C(C)O)C#N)C(F)(F)F |
| IUPAC-Name | 2-cyano-3-hydroxy-N-[2,3,5,6-tetradeuterio-4-(trifluoromethyl)phenyl]but-2-enamide |
| Molekulargewicht (g/mol) | 274.23 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?