Learn More
Testosteron-2,2,4,6,6-d5-Undecanoat, CDN
Specifications
Specifications
| Chemischer Name oder Material | Testosterone-2,2,4,6,6-D5 17-Undecanoate |
| Summenformel | C30 D5 H43 O3 |
| Menge | 0.005 g |
| Synonym | Testosterone-2,2,4,6,6-D5 17-undecanoate, Testosterone-2,2,4,6,6-d5 undecanoate, (17β)-17-[(1-Oxoundecyl)oxy]androst-4-en-3-one-2,2,4,6,6-d5, Testosterone-2,2,4,6,6-d5 undecylate |
| InChI-Formel | InChI=1 S/C30H48O3/c1-4-5-6-7-8-9-10-11-12-28(32)33-27-16-15-25-24-14-13-22-21-23(31)17-19-29(22,2)26(24)18-20-30(25,27)3/h21,24-27 H,4-20H2,1-3H3/t24-,25-,26-,27-,29-,30-/m0/s1/i13D2,17D2,21 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@H]3 [C@@H]4 CC[C@H](OC(=O)CCCCCCCCCC)[C@@]4(C)CC[C@@H]3 [C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC-Name | [(8R,9S,10R,13S,14S,17S)-2,2,4,6,6-Pentadeuterio-10,13-dimethyl-3-oxo-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl]undecanoat |
| Molekulargewicht (g/mol) | 461.731 |
| Formelmasse | 461,392 g/mol |
| Reinheit (%) | 98 Atom % D, Minimum 98% chemische Reinheit |