Learn More
Invitrogen™ Testosterone Recombinant Rabbit Monoclonal Antibody (RM435)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA538676
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Description
Testosterone Recombinant Monoclonal Antibody for ELISA
Testosterone is a steroid hormone from the androgen group and is found in mammals, reptiles, birds, and other vertebrates. In mammals, testosterone is primarily secreted in the testes of males and the ovaries of females, although small amounts are also secreted by the adrenal glands. It is the principal male sex hormone and an anabolic steroid.
Specifications
| Testosterone | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| Rabbit | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA | |
| RM435 | |
| Unconjugated | |
| Androderm; C19H28O2; Mertestate; Sustanon; Testoderm; Testosteron | |
| Testosterone-KLH. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |