missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP3-11863SS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TET1 Polyclonal antibody specifically detects TET1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)
Spezifikation
| TET1 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) | |
| 1.0 mg/mL | |
| Western Blot 2 - 4 μg/ml, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1 - 2 μg/ml, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) 1 - 3 μg/ml | |
| bA119F7.1, CXXC finger 6, CXXC6, CXXC6FLJ10839, FLJ41442, KIAA1676CXXC-type zinc finger protein 6, LCX, LCXEC 1.14.11.n2, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, Ten-eleven translocation 1 gene protein, ten-eleven translocation-1, tet oncogene 1 | |
| Partial recombinant human TET1 protein produced in E. coli (amino acids 1-206) [UniProt Q8NFU7]. | |
| 0.025 mg | |
| Cancer, Epigenetics, Neuroscience | |
| 80312 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction