missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Tetanus Toxin Polyclonal antibody specifically detects Tetanus Toxin in Bacteria samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Tetanus Toxin |
| Anwendungen | ELISA |
| Klassifikation | Polyclonal |
| Konjugat | FITC |
| Zusammensetzung | PBS |
| Gen-Alias | Tentoxylysin |
| Wirtsspezies | Rabbit |
| Immunogen | Synthetic peptide corresponding to the amino acids 1300 - 1314 of the native molecule conjugated to Keyhole limpet Haemocyanin |
| Reinigungsverfahren | Immunogen affinity purified |
| Menge | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?