Learn More
Invitrogen™ TFE3 Recombinant Rabbit Monoclonal Antibody (24GB2630)
Rabbit Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA557771
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30275620
  • 204.00 € / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
TFE3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. This protein contains a bipartite nuclear localization signal. This gene is known to escape chromosome X-inactivation.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
TFE3
Recombinant Monoclonal
0.8 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P19532, Q64092
TFE3
A synthesized peptide derived from human TFE3 (aa 150-450).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2630
Unconjugated
TFE3
bHLHe33; Class E basic helix-loop-helix protein 33; F830016E06Rik; RCCP2; RCCX1; RGD1559642; Tcfe3; TFE3; Tfe-3; TFEA; transcription factor binding to IGHM enhancer 3; transcription factor E family, member A; transcription factor E3; transcription factor for IgH enhancer; transcription factor for immunoglobulin heavy-chain enhancer 3
Rabbit
Affinity chromatography
RUO
209446, 317376, 7030
-20°C