Learn More
TFEB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Artikelnummer. 16841544 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
150 μL
20 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
16841544 20 μL 20 Mikroliter
16831544 150 μL 150 Mikroliter
2 options
Artikelnummer. 16841544
Marke: Proteintech 133721AP20UL
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
This is a rabbit polyclonal antibody raised against the full length of human TFEB. The calculated molecular weight of TFEB is 53 kDa, but modified TFEB is about 65-70 kDa.
The Transcription factor EB (TFEB) is a member in the basic helix-loop-helix leucine zipper superfamily of transcription factors that is translocated in a subset of renal tumors. Recent studies have shown that lysosomal biogenesis is regulated by TFEB, which is in turn regulated by the mammalian target of rapamycin (mTOR) complex 1. Other evidence suggests that TFEB coordinates the major steps of the autophagic pathway by driving the expression of autophagy and lysosomal genes.
Spezifikation
Antigen TFEB
Anwendungen Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Klassifikation Polyclonal
Konzentration 0.35 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen TFEB
Gen-Zugriffsnummer P19484, Q9R210
Gen-Alias AlphaTFEB, bHLHe35, TCFEB, TFEB, transcription factor EB
Gensymbole Tfeb
Wirtsspezies Rabbit
Immunogen TFEB Fusion Protein Ag4194
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 21425, 316214, 7942
Zielspezies Human, Rat, Mouse
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG