missing translation for 'onlineSavingsMsg'
Learn More

TFF1/pS2 Mouse, Clone: SPM573, Novus Biologicals™

Artikelnummer. 18145013 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.02 mg
0.1 mg
Packungsgröße:
0.02 Milligramm
0.10 Milligramm
Artikelnummer. Menge unitSize
18145013 0.02 mg 0.02 Milligramm
18115964 0.1 mg 0.10 Milligramm
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 18145013

Marke: Novus Biologicals NBP2329000.02MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

TFF1/pS2 Monoclonal specifically detects TFF1/pS2 in Human, Cynomolgus Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen TFF1/pS2
Anwendungen Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon SPM573
Konjugat Unconjugated
Verdünnung Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 0.5-1.0ug/ml
Zusammensetzung PBS with 0.05% BSA. with 0.05% Sodium Azide
Gen-Zugriffsnummer P04155
Gen-Alias BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A
Gensymbole TFF1
Wirtsspezies Mouse
Immunogen Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the C-terminus of human pS2.
Molekulargewicht des Antigens 6.5 kDa
Reinigungsverfahren Protein A purified
Menge 0.02 mg
Regulatorischer Status RUO
Forschungsgebiet Breast Cancer, Cancer
Primär oder sekundär Primary
Gen-ID (Entrez) 7031
Testspezifität It recognizes a polypeptide of 6.5kDa, identified as pS2 estrogen-regulated protein. Its epitope is localized between aa57-84 of human pS2 protein. pS2 is a trefoil peptide. Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing. These peptides contain three intra-chain disulfide bonds, forming the trefoil motif, or P-domain. pS2 is known to form dimers and this dimerization is thought to play a role in its protective and healing properties. About 60% of breast carcinomas are positive for pS2. Staining is cytoplasmic, often with localization to the Golgi apparatus. pS2 is shown to be localized in normal stomach mucosa, gastric fluid, goblet cells in the colon and small intestine, and in ulcerations of the gastrointestinal tract. Several studies have shown that pS2 is primarily expressed in estrogen receptor-positive breast tumors and it may define a subset of estrogen-dependent tumors that displays an increased likelihood of response to endocrine therapy.
Zielspezies Human, Primate
Inhalt und Lagerung Store at 4C.
Form Purified
Isotype IgG1 κ
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.