Learn More
TFPI Monoclonal Antibody (OTI3B1), TrueMAB™, OriGene
Beschreibung
TFPI Monoclonal antibody specifically detects TFPI in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Spezifikation
Spezifikation
| Antigen | TFPI |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI3B1 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gen | Angptl1 |
| Gen-Zugriffsnummer | O95841 |
| Gen-Alias | A630013F22Rik; anti-convertin; AW552122; EPI; Extrinsic pathway inhibitor; LACI; Lipoprotein-associated coagulation inhibitor; OTTHUMP00000163472; OTTHUMP00000205431; TFI; TFPI; TFPI1; Tissue factor pathway inhibitor; tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) |
| Mehr anzeigen |